Learn More
Masterflex™ PFA Threaded Adapter Barbed Fitting
Product Code. 11719337
Click to view available options
For Use With:
1/4 NPTM x 3/8 in. tubing I.D.
3/8 NPTM x 3/8 in. tubing I.D.
Thread Size:
1/4 in. NPTM
3/8 in. NPTM
Unit Size:
Each
Product Code. 11719337
Brand: Masterflex™ 0638394
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Providing quick and easy assembly when connecting a NPT male thread to a hose barb.
  • Connect fluid paths without the use of tools
  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • No leaks, flow restrictions or costly downtime
  • Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PFA
For Use With 3/8 NPTM x 3/8 in. tubing I.D.
Packaging Quantity 1 Each
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 3/8 in. NPTM
Product Type Barbed Fitting