Learn More
Cole-Parmer™ PFA Threaded Plug
Click to view available options
Thread Size:
1/2 in.
3/8 in.
Unit Size:
Each
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
| Type | Plug |
| Color | Clear |
| Thread Size | 3/8 in. |
| Port Type | Male NPT Fitting |
| Sterilization Method | Ethylene Oxide (ETO) Only |
| Overall Length (English) | 27/32 in. |
| Material | PFA |
| Overall Width (English) | 13/16 in. |