missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN1 Antibody (12), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06609UV
This item is not returnable.
View return policy
Description
PFDN1 Monoclonal antibody specifically detects PFDN1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PFDN1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 12 | |
| Western Blot, ELISA | |
| PDF, PFD1prefoldin 1, prefoldin subunit 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN1 (NP_002613.2; Met1-Gln122). | |
| 0.1 mL | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 5201 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction