missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN1 Antibody (12), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | PFDN1 |
|---|---|
| Clone | 12 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643387
|
Novus Biologicals
NBP3-06609-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633197
|
Novus Biologicals
NBP3-06609-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PFDN1 Monoclonal antibody specifically detects PFDN1 in Human samples. It is validated for Western Blot, ELISASpecifications
| PFDN1 | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| PDF, PFD1prefoldin 1, prefoldin subunit 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN1 (Accession#: NP_002613.2; Met1-Gln122). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 0.2 um filtered solution in PBS | |
| 5201 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title