missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PFDN1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142101
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.
Specifications
| PFDN1 | |
| Polyclonal | |
| Unconjugated | |
| PFDN1 | |
| 2700086I23Rik; AA408327; AU044714; PDF; PFD1; PFDN1; prefoldin 1; prefoldin subunit 1; Unknown (protein for MGC:128261) | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60925 | |
| PFDN1 | |
| Peptide with sequence C-DNIREMLMARRAQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction