missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN2 Antibody (04), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 326.00
Specifications
| Antigen | PFDN2 |
|---|---|
| Clone | 04 |
| Dilution | Immunocytochemistry/ Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647598
|
Novus Biologicals
NBP3-06549-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637368
|
Novus Biologicals
NBP3-06549-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PFDN2 Monoclonal antibody specifically detects PFDN2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| PFDN2 | |
| Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| PFD2, prefoldin 2, prefoldin subunit 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN2 (Uniprot#: Q9UHV9; Met1-Ser154). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 04 | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5202 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title