missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN2 Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06550-100ul
This item is not returnable.
View return policy
Description
PFDN2 Monoclonal antibody specifically detects PFDN2 in Human samples. It is validated for ELISA
Specifications
| PFDN2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 05 | |
| ELISA 1:1000-1:2000 | |
| PFD2, prefoldin 2, prefoldin subunit 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN2 (Uniprot#: Q9UHV9; Met1-Ser154). | |
| 100 μg | |
| Signal Transduction | |
| 5202 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction