Learn More
PFDN2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00279JF549
Product Code. 18580030
  • € 407.00 / 0.10mL
Description
PFDN2 Polyclonal antibody specifically detects PFDN2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PFDN2
Western Blot, ELISA, Immunoprecipitation
PFD2, prefoldin 2, prefoldin subunit 2
Janelia Fluor 549
50mM Sodium Borate
5202