missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PFDN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580890
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PFD2/PFDN2.
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
Specifications
| PFDN2 | |
| Polyclonal | |
| Unconjugated | |
| PFDN2 | |
| ESTM27; HSPC231; PFD2; Pfdn2; prefoldin 2; prefoldin subunit 2; similar to prefoldin subunit 2; W48336 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9UHV9 | |
| PFDN2 | |
| Recombinant Human PFD2/PFDN2 protein (Met1-Ser154). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction