missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN5 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97061IR
This item is not returnable.
View return policy
Description
PFDN5 Polyclonal specifically detects PFDN5 in Human samples. It is validated for ELISA.
Specifications
| PFDN5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5204 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 755 | |
| c-myc binding protein, C-Myc-binding protein Mm-1, MM-1, MM1MGC71907, Myc modulator 1, myc modulator-1, PFD5MGC5329, prefoldin 5, prefoldin subunit 5 | |
| Produced in rabbits immunized with purified, recombinant PFDN5 (NP_002615.2; Met1-Ala154). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction