missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN6 Antibody (OTI4E10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73339F
This item is not returnable.
View return policy
Description
PFDN6 Monoclonal specifically detects PFDN6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| PFDN6 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PFDN6 | |
| Recombinant protein fragment corresponding to amino acids 2-129 of human PFDN6 (NP_055075) produced in E.coli. | |
| 0.1 mL | |
| 10471.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry | |
| OTI4E10 | |
| Flow Cytometry, Immunohistochemistry | |
| H2-KE2, HLA class II region expressed gene KE2, KE-2, KE2 protein, MGC70744, PFD6HKE2, prefoldin 6, prefoldin subunit 6, Protein Ke2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction