missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN6 Antibody (OTI4E10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73339MFV500
This item is not returnable.
View return policy
Description
PFDN6 Monoclonal antibody specifically detects PFDN6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| PFDN6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI4E10 | |
| Flow Cytometry | |
| H2-KE2, HLA class II region expressed gene KE2, KE-2, KE2 protein, MGC70744, PFD6HKE2, prefoldin 6, prefoldin subunit 6, Protein Ke2 | |
| Human recombinant protein fragment corresponding to amino acids 2-129 of human PFDN6(NP_055075) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 10471 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction