missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFK-158, MedChemExpress
Description
PFK-158 is a potent and selective PFKFB3 inhibitor with an IC50 value 137 nM. PFK-158 reduces glucose uptake, ATP production, lactate release, and induces apoptosis and autophagy in cancer cells. PFK-158 has broad anti-tumor activity. PFK-158 can also enhance Colistin's resistance to bacteria.
Specifications
Specifications
| Chemical Name or Material | PFK-158 |
| Physical Form | Solid |
| CAS | 1462249-75-7 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H11F3N2O |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 30 mg/mL (91.38 mM) |
| SMILES | O=C(C1=CC=NC=C1)/C=C/C2=NC3=CC(C(F)(F)F)=CC=C3C=C2 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?