Learn More
PFKFB2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32194DL594
Product Code. 18719821
  • € 669.00 / 0.10mL
Description
PFKFB2 Polyclonal antibody specifically detects PFKFB2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PFKFB2
RUO
5208