missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB4 Antibody (OTI1C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02226
This item is not returnable.
View return policy
Description
PFKFB4 Monoclonal specifically detects PFKFB4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PFKFB4 | |
| Monoclonal | |
| 0.9 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| PFKFB4 | |
| Full length human recombinant protein of human PFKFB4 (NP_004558) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 5210 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1C8 | |
| Unconjugated | |
| 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4,6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4,6PF-2-K/Fru-2,6-P2ase 4,6PF-2-K/Fru-2,6-P2ase testis-type isozyme, bifunctional enzyme with kinase and biphosphatase activities, PFK/FBPase 4 | |
| Mouse | |
| 54 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction