missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | PFKFB4 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The PFKFB4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PFKFB4. This antibody recognizes human, mouse antigen. The PFKFB4 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| PFKFB4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 5210 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title