missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKL, Mouse anti-Human, Clone: 2A9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005211-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: PKEKSNFSLAILNVGAPAAGMNAAVRSAVRTGISHGHTVYVVHDGFEGLAKGQVQEVGWHDVAGWLGRGGSMLGTKRTLPKGQLESIVENIRIYGIHALLSpecifications
| PFKL | |
| Monoclonal | |
| Unconjugated | |
| phosphofructokinase, liver | |
| DKFZp686G1648/DKFZp686L2097/FLJ30173/FLJ40909/PFK-B | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A9 | |
| In 1x PBS, pH 7.4 | |
| NM_001002021 | |
| PFKL | |
| PFKL (NP_001002021, 441 a.a. ∼ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5211 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction