missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKP Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99101G
This item is not returnable.
View return policy
Description
PFKP Polyclonal antibody specifically detects PFKP in Human samples. It is validated for Western Blot
Specifications
| PFKP | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PFKP. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5214 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction