missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKP Antibody (OTI1D6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73344F
This item is not returnable.
View return policy
Description
PFKP Monoclonal specifically detects PFKP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| PFKP | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PFKP | |
| Full length recombinant protein of human PFKP (NP_002618) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| OTI1D6 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| 6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 5214.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction