Learn More
PFN1 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI1D5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501211S
Encompass_Preferred
Product Code. 17583601
  • € 242.00 / 30µL
Description
Profilin is an ubiquitous small (12-15 kDa) phosphoinositides and poly-L-proline binding protein that plays a role in both signal transduction pathways and actin filament dynamics. There are two mammalian profilins with similar biochemical properties but different expression patterns. Profilin I appears to be highly expressed in most tissues except for skeletal muscle. Profilin II is predomitly expressed in brain and at lower levels in skeletal muscle, uterus and kidney. Profilin is a mainly cytosolic protein with higher concentrations in dynamic membrane areas like the leading edge and ruffling membranes.
Specifications
Specifications
PFN1
Monoclonal
Unconjugated
PFN1
ALS18
Mouse
Affinity Chromatography
RUO
5216, 607397, 64303
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
OTI1D5
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P07737, P62963
Pfn1
Full length human recombit protein of human PFN1 produced in HEK293T cell.
30 μL
Primary
Canine, Human, Non-human Primate, Rat
Antibody
IgG2a