missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGA5 Monoclonal Antibody (2C1)

Description
MA5-15558 targets PGA5 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15558 immunogen is purified recombinant fragment of human PGA5 expressed in E. Coli. MA5-15558 detects PGA5 which has a predicted molecular weight of approximately 42kDa.
PGA5: Pepsinogen 5, group I (pepsinogen A). Pepsinogens are the inactive precursors of pepsin, the major acid protease found in the stomach. Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. PGA5 is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. PGA5 is inhibited by ovUS-1, a uterine serpin.
Specifications
Specifications
| Antigen | PGA5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2C1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | Pga5 |
| Gene Accession No. | P0DJD9 |
| Gene Alias | 1110035E17Rik; Pepf; pepsin A; pepsin A-5; Pepsin F; Pepsinogen 5; pepsinogen 5, group I; pepsinogen 5, group I (pepsinogen A); pepsinogen A; pepsinogen A precursor; pepsinogen A5; pepsinogen F; pepsinogen F protein; pepsinogen precursor; pepsinogen-5; Pg5; PGA; PGA3; PGA4; PGA5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?