missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97513AF750
This item is not returnable.
View return policy
Description
PGAM1 Polyclonal specifically detects PGAM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PGAM1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B | |
| Produced in rabbits immunized with E. coli-derived Human PGAM1 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5223 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction