Learn More
PGAM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16126-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PGAM1.
PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombit human PGAM protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.Specifications
| PGAM1 | |
| Polyclonal | |
| Unconjugated | |
| PGAM1 | |
| BPG dependent PGAM 1, PGAM B, PGAM1, PGAMA, Phosphoglycerate mutase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18648, 24642, 5223 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18669, P25113, Q9DBJ1 | |
| PGAM1 | |
| PGAM1 Fusion Protein Ag9110 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |