missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM2 Antibody (OTI4E9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73358PEATT594
This item is not returnable.
View return policy
Description
PGAM2 Monoclonal specifically detects PGAM2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications
| PGAM2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| PGAM2 | |
| Full length recombinant protein of human PGAM2 (NP_000028) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG2a |
| Flow Cytometry | |
| OTI4E9 | |
| Flow Cytometry | |
| BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
| Mouse | |
| Affinity Purified | |
| Cancer, Proteases & Other Enzymes, Signal Transduction | |
| 5224.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction