missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGBD3 Monoclonal Antibody (OTI3H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810308S
This item is not returnable.
View return policy
Description
PGBD3 Monoclonal antibody specifically detects PGBD3 in Human samples. It is validated for Western BlotSpecifications
| PGBD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50458 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3H1 | |
| Unconjugated | |
| Lhx2 | |
| 4732403I04; ATP-dependent helicase ERCC6; C130058G22Rik; Cockayne syndrome protein CSB; CS group B correcting; Csb; DNA excision repair protein ERCC-6; Ercc6; excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; FLJ90201; PGBD3; piggyBac transposable element derived 3; piggyBac transposable element-derived protein 3 | |
| Human recombinant protein fragment corresponding to amino acids 220-406 of human LHX2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction