missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGBD3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA541302

Product Code. 15524762

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: NLPGSLLHTA AYLIQDGSDA ESDSDDPSYA PKDDSPDEVP STFTVQQPPP Sequence homology: Human: 100%.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. Anti-PGBD3 belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. Anti-PGBD3 overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12. The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PGBD3
Polyclonal
Unconjugated
PGBD3
4732403I04; ATP-dependent helicase ERCC6; C130058G22Rik; Cockayne syndrome protein CSB; CS group B correcting; Csb; DNA excision repair protein ERCC-6; Ercc6; excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; FLJ90201; PGBD3; piggyBac transposable element derived 3; piggyBac transposable element-derived protein 3
Rabbit
Protein A
RUO
267004
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8N328
PGBD3
Synthetic peptide directed towards the N-terminal of human PGBD3 (aa 52-101).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.