missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGBD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141035
This item is not returnable.
View return policy
Description
PGBD5 Polyclonal Antibody for Western Blot, IHC (P)
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families.
Specifications
| PGBD5 | |
| Polyclonal | |
| Unconjugated | |
| PGBD5 | |
| 2900019M05Rik; AI854313; DKFZp761A0620; FLJ11413; PGBD5; piggyBac domain related protein 5; PiggyBac domain-related protein 5; piggyBac transposable element derived 5; piggyBac transposable element-derived protein 5; PiggyBac transposase 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 209966, 292098, 79605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8N414 | |
| PGBD5 | |
| Recombinant protein within human PGBD5 aa 270-520. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction