missing translation for 'onlineSavingsMsg'
Learn More

PGC, Mouse anti-Human, Clone: L10CC10, Abnova™

Product Code. 16056766
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16056766 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16056766 Supplier Abnova Supplier No. MAB5251.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen PGC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone L10CC10
Conjugate Unconjugated
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene progastricsin (pepsinogen C)
Gene Alias -
Gene Symbols PGC
Host Species Mouse
Immunogen Native purified human PGC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5225
Test Specificity This antibody is specific to human pepsinogen II with no crossreactivity to human pepsinogen I. Pepsinogen II is a group of precursor molecules for pepsin. These proteins are secreted into the gastric lumen by the pyloric glands of the gastric antrum and also by the chief and neck cells of the gastric corpus (oxyntic mucosa).
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.