Learn More
PGC1a Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Product Code. 16838185 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16838185 20 μL 20µL
16828185 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16838185 Supplier Proteintech Supplier No. 663691IG20UL
Mouse Monoclonal Antibody
The PGC1a antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human PGC1a. This antibody recognizes human, mouse, rat antigen. The PGC1a antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Antigen PGC1a
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1C1B2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 10891
Target Species Human, Rat, Mouse
Isotype IgG1