missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGCD Polyclonal Antibody

Description
PGCD Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Probably plays a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction.
Specifications
Specifications
| Antigen | PGCD |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | GUCY2D |
| Gene Accession No. | Q02846 |
| Gene Alias | cone rod dystrophy 6; CORD5; CORD6; CYGD; guanylate cyclase 2d; guanylate cyclase 2D, membrane (retina-specific); guanylate cyclase 2D, retinal; Guanylate cyclase, olfactory; guanylyl cyclase 2d; guanylyl cyclase D; GUC1A4; GUC2D; GUCY2D; LCA; LCA1; olfactory guanylyl cyclase GC-D; RCD2; RETGC; RETGC1; RETGC-1; retinal guanylate cyclase 1; retinal guanylyl cyclase 1; Rod outer segment membrane guanylate cyclase; ROSGC; ROS-GC; ROS-GC1 |
| Gene Symbols | GUCY2D |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?