missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD Antibody (OTI2A5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73361APC
This item is not returnable.
View return policy
Description
PGD Monoclonal antibody specifically detects PGD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| PGD | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI2A5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| 6PGD, EC 1.1.1.44,6-phosphogluconate dehydrogenase, decarboxylating, PGDH, phosphogluconate dehydrogenase | |
| Full length human recombinant protein of human PGD(NP_002622) produced in HEK293T cell. | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 5226 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction