missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGD Monoclonal Antibody (OTI2A5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526165
This item is not returnable.
View return policy
Description
PGD Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
Specifications
| PGD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52209 | |
| PGD | |
| Full length human recombinant protein of PGD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2A5 | |
| Unconjugated | |
| PGD | |
| 0610042A05Rik; 6PGD; 6-phosphogluconate dehydrogenase, decarboxylating; 6-phosphogluconate dehydrogenase, decarboxylating-like; AU019875; C78335; LOC100360180; Pgd; PGDH; phosphogluconate dehydrogenase | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5226 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction