missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGD Polyclonal Antibody
GREENER_CHOICE

Product Code. 13401597
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13401597 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13401597 Supplier Invitrogen™ Supplier No. PA527486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, Mouse heart, Mouse lung. Predicted reactivity: Mouse (91%), Rat (90%), Zebrafish (86%), Xenopus laevis (84%), Chicken (91%), Sheep (96%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PGD
Gene Accession No. P52209, Q9DCD0
Gene Alias 0610042A05Rik; 6PGD; 6-phosphogluconate dehydrogenase, decarboxylating; 6-phosphogluconate dehydrogenase, decarboxylating-like; AU019875; C78335; LOC100360180; Pgd; PGDH; phosphogluconate dehydrogenase
Gene Symbols PGD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PGD. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110208, 5226
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.