missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGF Recombinant Rabbit Monoclonal Antibody (6N4V2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542783
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse heart. Immunogen sequence: MPVMRLFPCF LQLLAGLALP AVPPQQWALS AGNGSSEVEV VPFQEVWGRS YCRALERLVD VVSEYPSEVE HMFSPSCVSL LRCTGCCGDE NLHCVPVETA.
Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
Specifications
| PGF | |
| Recombinant Monoclonal | |
| 0.55 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49763, P49764, Q63434 | |
| PGF | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6N4V2 | |
| Unconjugated | |
| PGF | |
| AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18654, 5228, 94203 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction