missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGK1/PGK2 Recombinant Rabbit Monoclonal Antibody (HL1775)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547177
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The protein encoded by this gene is a glycolytic enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate The encoded protein may also act as a cofactor for polymerase alpha Additionally, this protein is secreted by tumor cells where it participates in angiogenesis by functioning to reduce disulfide bonds in the serine protease, plasmin, which consequently leads to the release of the tumor blood vessel inhibitor angiostatin The encoded protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions Deficiency of the enzyme is associated with a wide range of clinical phenotypes hemolytic anemia and neurological impairment Pseudogenes of this gene have been defined on chromosomes 19, 21 and the X chromosome.
Specifications
| PGK1/PGK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| HEL-S-68p; MIG10; PGKA; phosphoglycerate kinase 1 | |
| Recombinant protein of human PGK1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1775 | |
| Unconjugated | |
| P29452, P29466, P43527, P48729, P97633, Q8BK63, Q8N752 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 0, 113927, 122011, 12362, 1452, 834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction