missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGK2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99497V
This item is not returnable.
View return policy
Description
PGK2 Polyclonal antibody specifically detects PGK2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PGK2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| DyLight 405 | |
| dJ417L20.2, EC 2.7.2.3, PGK-2, PGKB, PGKPS, phosphoglycerate kinase 2, phosphoglycerate kinase autosomal pseudogene, Phosphoglycerate kinase, testis specific | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PGK2. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 5232 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction