missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118159
This item is not returnable.
View return policy
Description
PGK2 Polyclonal Antibody for Western Blot, IP
PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.
Specifications
| PGK2 | |
| Polyclonal | |
| PBS with no preservative | |
| P07205 | |
| PGK2 | |
| A synthetic peptide corresponding to the center region of the Human PGK2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PGK2 | |
| dJ417L20.2; epididymis secretory protein Li 272; HEL-S-272; Pgk2; Pgk-2; PGKB; PGKPS; phosphoglycerate kinase 2; phosphoglycerate kinase autosomal pseudogene; phosphoglycerate kinase, testis specific; Tcp-2; testicular tissue protein Li 139 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5232 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction