Learn More
PGK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13686-1-AP-150UL
 View more versions of this product
Product Code. 16852034
  • € 410.00 / 150µL
Description
PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.
Specifications
Specifications
PGK2
Polyclonal
Unconjugated
PGK2
PGK 2, PGK2, PGKB, PGKPS, phosphoglycerate kinase 2
Rabbit
Antigen Affinity Chromatography
RUO
18663, 316265, 5232
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07205, P09041
Pgk2
PGK2 Fusion Protein Ag4553
150 μL
Primary
Human, Mouse, Rat