missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13686-1-AP-20UL
This item is not returnable.
View return policy
Description
PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.Specifications
| PGK2 | |
| Polyclonal | |
| Unconjugated | |
| PGK2 | |
| PGK 2, PGK2, PGKB, PGKPS, phosphoglycerate kinase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18663, 316265, 5232 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07205, P09041 | |
| Pgk2 | |
| PGK2 Fusion Protein Ag4553 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction