Learn More
PGLYRP1/PGRP-S Antibody (188C424), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56719PECY7
This item is not returnable.
View return policy
Description
PGLYRP1/PGRP-S Monoclonal specifically detects PGLYRP1/PGRP-S in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| PGLYRP1/PGRP-S | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| PGLYRP1 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG3 κ |
| Flow Cytometry | |
| 188C424 | |
| Flow Cytometry | |
| peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L | |
| Mouse | |
| Protein G purified | |
| Apoptosis | |
| 8993.0 | |
| Store at 4°C in the dark. Do not freeze. |