missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PGLYRP1/PGRP-S Antibody (Novo Nordisk patent anti-PGLYRP1) - Humanized, IgG4SP, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PGLYRP1/PGRP-S Monoclonal antibody specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | PGLYRP1/PGRP-S |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Novo Nordisk patent anti-PGLYRP1 |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | O75594 |
| Gene Alias | peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L |
| Host Species | Human |
| Immunogen | PGLYRP1/ PGRP-S |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?