missing translation for 'onlineSavingsMsg'
Learn More

PGLYRP1/PGRP-S Antibody (Novo Nordisk patent anti-PGLYRP1) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30177984
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30177984 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30177984 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328593JF635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

PGLYRP1/PGRP-S Monoclonal antibody specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGLYRP1/PGRP-S
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Novo Nordisk patent anti-PGLYRP1
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Host Species Human
Immunogen PGLYRP1/ PGRP-S
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8993
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.