Learn More
PGLYRP1/PGRP-S Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21331-25ul
 View more versions of this product
Product Code. 18649694
  • € 450.00 / 25µL
Description
PGLYRP1/PGRP-S Polyclonal antibody specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PGLYRP1/PGRP-S
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
This antibody was developed against Recombinant Protein corresponding to amino acids: VPRNEWKALASECAQHLSLPLRYVVVSHTAGSSCNTPASCQQQARNVQHYHMKTLGWCDVGYNF
25 μg
Apoptosis
8993
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified