missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP2/PGRP-L Antibody (439728), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4704K
This item is not returnable.
View return policy
Description
PGLYRP2/PGRP-L Monoclonal antibody specifically detects PGLYRP2/PGRP-L in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| PGLYRP2/PGRP-L | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 439728 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPLHMFT0141, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like | |
| Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L, Ser23-Asn530, Accession # Q8VCS0 | |
| 0.1 mL | |
| Immunology, Inflammation | |
| 114770 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction