missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP4/PGRP-I beta Antibody (186C426), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56721FR
This item is not returnable.
View return policy
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).
Specifications
| PGLYRP4/PGRP-I beta | |
| Monoclonal | |
| DyLight 680 | |
| Q96LB8 | |
| PGLYRP4 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta. | |
| 0.1 mL | |
| Primary | |
| PGRP-1b (186C426) | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| 186C426 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular) | |
| peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57115 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction