Learn More
PGM1 Antibody (CL3299) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44049
Product Code. 30301963
  • € 638.00 / 100µL
Description
PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PGM1
Monoclonal
LYOPH
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000
EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1
This antibody was generated using a recombinant protein sequence of P36871, with the exact immunogen sequence remaining proprietary.
100 μL
Lipid and Metabolism
5236
Human
Purified
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
CL3299
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.