missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM2, Mouse anti-Human, Clone: 1A3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00055276-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAPEGSGLGEDARLDQETAQWLRWDKNSLTLEAVKRLIAEGNKEELRKCFGARMEFGTAGLRAAMGPGISRMNDLTIIQTTQGFCRYLESpecifications
| PGM2 | |
| Monoclonal | |
| Unconjugated | |
| phosphoglucomutase 2 | |
| FLJ10983/MSTP006 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A3 | |
| In 1x PBS, pH 7.4 | |
| BC010087.2 | |
| PGM2 | |
| PGM2 (AAH10087.1, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55276 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur