missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118160
This item is not returnable.
View return policy
Description
PGM2 Polyclonal Antibody for Western Blot, IP
Catalyzes the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses. May also catalyze the interconversion of glucose-1-phosphate and glucose-6-phosphate. Has low glucose 1,6-bisphosphate synthase activity. [UniProt].
Specifications
| PGM2 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96G03 | |
| PGM2 | |
| E. coli-derived Human PGM2/phosphoglucomutase 2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PGM2 | |
| Glucose phosphomutase 2; MSTP006; PGM 2; PGM2; Phosphodeoxyribomutase; phosphoglucomutase 2; phosphoglucomutase-2; Phosphopentomutase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55276 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction