missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM2L1 Monoclonal Antibody (OTI4G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812136
This item is not returnable.
View return policy
Description
PGM2L1 is the Glucose 1, 6-bisphosphate synthase using 1, 3-bisphosphoglycerate as a phosphate donor and a series of 1-phosphate sugars as acceptors, including glucose 1-phosphate, mannose 1-phosphate, ribose 1-phosphate and deoxyribose 1-phosphate. 5 or 6-phosphosugars are bad substrates, with the exception of glucose 6-phosphate. PGM2L1 also synthesizes ribose 1, 5-bisphosphate. PGM2L1 has only low phosphopentomutase and phosphoglucomutase activities.Specifications
| PGM2L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6PCE3 | |
| PGM2L1 | |
| Full length human recombit protein of human PGM2L1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G6 | |
| Unconjugated | |
| PGM2L1 | |
| BM32A, PMMLP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 283209 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction