Learn More
PGM3 Mouse anti-Human, Clone: OTI1B8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504860
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.Specifications
| PGM3 | |
| Monoclonal | |
| Unconjugated | |
| PGM3 | |
| AGM1, IMD23, PAGM, PGM 3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5238 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95394 | |
| PGM3 | |
| Full length human recombit protein of human PGM3 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.